Assay Details
| Gene Symbol: Dsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144731.3 | NP_001138203.1 | 3 - 4 | 885 | 60 | |
| NM_001298393.1 | NP_001285322.1 | 5 - 6 | 926 | 60 | ||
| NM_080715.4 | NP_542446.2 | 5 - 6 | 1519 | 60 | ||
| NM_167502.3 | NP_727959.2 | 4 - 5 | 1583 | 60 | ||
| NM_167503.3 | NP_727960.1 | 5 - 6 | 998 | 60 | ||
| NM_167504.2 | NP_727961.1 | 5 - 6 | 923 | 60 | ||
| NM_206762.3 | NP_996485.1 | 5 - 6 | 1067 | 60 | ||
| GenBank mRNA | AY060841.1 | - | 5 - 6 | 910 | 60 | |
| BT126193.1 | - | 5 - 6 | 925 | 60 | ||
More Information
Set Membership: |
|

Dsp1
Genomic Map