Assay Details
| Gene Symbol: bves | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272823.1 | NP_001259752.1 | 2 - 3 | 1261 | 62 | |
| NM_001272824.1 | NP_001259753.1 | 2 - 3 | 1261 | 62 | ||
| NM_134582.4 | NP_608426.1 | 2 - 3 | 1424 | 62 | ||
| NM_206800.3 | NP_996523.1 | 2 - 3 | 1261 | 62 | ||
| GenBank mRNA | AF247183.1 | - | 2 - 3 | 1226 | 62 | |
| AY113513.1 | - | 2 - 3 | 1380 | 62 | ||
| BT132753.1 | - | 2 - 3 | 1241 | 62 | ||
More Information
Set Membership: |
|

bves
Genomic Map