Assay Details
| Gene Symbol: Npc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134573.3 | NP_608417.2 | 1 - 2 | 1266 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npc1b| Gene Symbol: Npc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134573.3 | NP_608417.2 | 1 - 2 | 1266 | 62 | |
Set Membership: |
|