Assay Details
| Gene Symbol: r-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134575.2 | NP_608419.1 | 2 - 3 | 1396 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

r-cup| Gene Symbol: r-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134575.2 | NP_608419.1 | 2 - 3 | 1396 | 73 | |
Set Membership: |
|