Assay Details
| Gene Symbol: CG15445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272827.1 | NP_001259756.1 | 2 - 3 | 406 | 67 | |
| NM_134592.4 | NP_608436.4 | 2 - 3 | 400 | 67 | ||
| NM_167729.2 | NP_728390.1 | 1 - 2 | 460 | 67 | ||
| NM_167730.2 | NP_728391.3 | 2 - 3 | 411 | 67 | ||
| NM_167731.2 | NP_728392.2 | 2 - 3 | 407 | 67 | ||
| GenBank mRNA | BT004870.1 | - | 1 - 2 | 438 | 67 | |
| BT128735.1 | - | 2 - 3 | 404 | 67 | ||
More Information
Set Membership: |
|

CG15445
Genomic Map