Assay Details
| Gene Symbol: CG14621 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298593.1 | NP_001285522.1 | 1 - 2 | 1082 | 68 | |
| NM_001298594.1 | NP_001285523.1 | 2 - 3 | 587 | 68 | ||
| NM_001298595.1 | NP_001285524.1 | 2 - 3 | 475 | 68 | ||
| NM_134614.2 | NP_608458.1 | 1 - 2 | 560 | 68 | ||
| GenBank mRNA | BT003539.1 | - | 1 - 2 | 555 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001298595.1 transcripts.
Set Membership: |
|

CG14621
Genomic Map