Assay Details
| Gene Symbol: CG14621 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298593.1 | NP_001285522.1 | 3 - 4 | 1418 | 69 | |
| NM_001298594.1 | NP_001285523.1 | 4 - 5 | 923 | 69 | ||
| NM_001298595.1 | NP_001285524.1 | 4 - 5 | 811 | 69 | ||
| NM_134614.2 | NP_608458.1 | 3 - 4 | 896 | 69 | ||
| GenBank mRNA | BT003539.1 | - | 3 - 4 | 891 | 69 | |
More Information
Set Membership: |
|

CG14621
Genomic Map