Assay Details
| Gene Symbol: cbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272903.1 | NP_001259832.1 | 2 | 2080 | 126 | |
| NM_134685.3 | NP_608529.2 | 1 | 2962 | 126 | ||
| NM_164384.2 | NP_722636.1 | 2 | 1760 | 126 | ||
| GenBank mRNA | BT025196.1 | - | 1 | 2320 | 126 | |
| BT099918.1 | - | 2 | 1754 | 126 | ||
More Information
Set Membership: |
|

cbt
Genomic Map