Assay Details
| Gene Symbol: Saf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134701.4 | NP_608545.1 | 1 - 2 | 473 | 69 | |
| GenBank mRNA | AY069583.1 | - | 1 - 2 | 444 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Saf6| Gene Symbol: Saf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134701.4 | NP_608545.1 | 1 - 2 | 473 | 69 | |
| GenBank mRNA | AY069583.1 | - | 1 - 2 | 444 | 69 | |
Set Membership: |
|