Assay Details
| Gene Symbol: Saf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272914.1 | NP_001259843.1 | 4 - 5 | 1828 | 65 | |
| NM_134701.4 | NP_608545.1 | 4 - 5 | 1837 | 65 | ||
| GenBank mRNA | AY069583.1 | - | 4 - 5 | 1808 | 65 | |
More Information
Set Membership: |
|

Saf6
Genomic Map