Assay Details
| Gene Symbol: PNUTS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014453.2 | NP_001014453.1 | 2 - 3 | 1206 | 137 | |
| NM_001014454.2 | NP_001014454.1 | 3 - 4 | 510 | 137 | ||
| NM_001014455.2 | NP_001014455.1 | 3 - 4 | 418 | 137 | ||
| NM_001272921.1 | NP_001259850.1 | 3 - 4 | 418 | 137 | ||
| GenBank mRNA | AY061610.1 | - | 3 - 4 | 401 | 137 | |
More Information
Set Membership: |
|

PNUTS
Genomic Map