Assay Details
| Gene Symbol: mtRNApol | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272924.1 | NP_001259853.1 | 6 - 7 | 2922 | 62 | |
| NM_134721.3 | NP_608565.1 | 6 - 7 | 2922 | 62 | ||
| GenBank mRNA | BT011381.1 | - | 6 - 7 | 2901 | 62 | |
More Information
Set Membership: |
|

mtRNApol
Genomic Map