Assay Details
| Gene Symbol: Rim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134771.4 | NP_608615.1 | 2 - 3 | 735 | 96 | |
| GenBank mRNA | AY058735.1 | - | 2 - 3 | 681 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rim2| Gene Symbol: Rim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134771.4 | NP_608615.1 | 2 - 3 | 735 | 96 | |
| GenBank mRNA | AY058735.1 | - | 2 - 3 | 681 | 96 | |
Set Membership: |
|