Assay Details
| Gene Symbol: Rim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258911.2 | NP_001245840.1 | 5 - 6 | 1192 | 71 | |
| NM_001258912.2 | NP_001245841.1 | 5 - 6 | 1148 | 71 | ||
| NM_134771.4 | NP_608615.1 | 5 - 6 | 1213 | 71 | ||
| GenBank mRNA | AY058735.1 | - | 5 - 6 | 1159 | 71 | |
More Information
Set Membership: |
|

Rim2
Genomic Map