Assay Details
| Gene Symbol: Axud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272999.1 | NP_001259928.1 | 2 - 3 | 2493 | 76 | |
| NM_001273000.1 | NP_001259929.1 | 2 - 3 | 2071 | 76 | ||
| NM_134829.3 | NP_608673.1 | 2 - 3 | 2320 | 76 | ||
| NM_164480.2 | NP_722800.1 | 2 - 3 | 1982 | 76 | ||
| GenBank mRNA | AF181640.1 | - | 2 - 3 | 2330 | 76 | |
More Information
Set Membership: |
|

Axud1
Genomic Map