Assay Details
| Gene Symbol: Snx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273033.1 | NP_001259962.1 | 2 - 3 | 305 | 71 | |
| NM_134865.3 | NP_608709.1 | 2 - 3 | 314 | 71 | ||
| GenBank mRNA | AY061149.1 | - | 2 - 3 | 335 | 71 | |
More Information
Set Membership: |
|

Snx21
Genomic Map