Assay Details
| Gene Symbol: aph-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273034.1 | NP_001259963.1 | 1 - 2 | 476 | 65 | |
| NM_134866.3 | NP_608710.1 | 1 - 2 | 476 | 65 | ||
| GenBank mRNA | AF508786.1 | - | 1 - 2 | 415 | 65 | |
| AY051658.1 | - | 1 - 2 | 423 | 65 | ||
More Information
Set Membership: |
|

aph-1
Genomic Map