Assay Details
| Gene Symbol: HINT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273035.1 | NP_001259964.1 | 2 - 3 | 206 | 92 | |
| NM_134867.4 | NP_608711.3 | 2 - 3 | 206 | 92 | ||
| NM_164500.2 | NP_722836.1 | 1 - 2 | 214 | 92 | ||
| GenBank mRNA | AY070643.1 | - | 2 - 3 | 203 | 92 | |
| AY071745.1 | - | 2 - 3 | 203 | 92 | ||
More Information
Set Membership: |
|

HINT1
Genomic Map