Assay Details
| Gene Symbol: CG43707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273076.1 | NP_001260005.1 | 16 - 17 | 5727 | 73 | |
| NM_001273078.1 | NP_001260007.1 | 15 - 16 | 5592 | 73 | ||
| NM_001273079.1 | NP_001260008.1 | 2 - 3 | 274 | 73 | ||
More Information
Set Membership: |
|

CG43707
Genomic Map