Assay Details
| Gene Symbol: CG43707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273074.1 | NP_001260003.1 | 15 - 16 | 6025 | 78 | |
| NM_001273075.1 | NP_001260004.1 | 15 - 16 | 6587 | 78 | ||
| NM_001273076.1 | NP_001260005.1 | 17 - 18 | 6145 | 78 | ||
| NM_001273077.2 | NP_001260006.1 | 15 - 16 | 6025 | 78 | ||
| NM_001273078.1 | NP_001260007.1 | 16 - 17 | 6010 | 78 | ||
| NM_001273079.1 | NP_001260008.1 | 3 - 4 | 692 | 78 | ||
| NM_134962.3 | NP_608806.3 | 10 - 11 | 4784 | 78 | ||
More Information
Set Membership: |
|

CG43707
Genomic Map