Assay Details
| Gene Symbol: CG43707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273074.1 | NP_001260003.1 | 17 - 18 | 6391 | 85 | |
| NM_001273075.1 | NP_001260004.1 | 17 - 18 | 6953 | 85 | ||
| NM_001273076.1 | NP_001260005.1 | 19 - 20 | 6511 | 85 | ||
| NM_001273077.2 | NP_001260006.1 | 17 - 18 | 6391 | 85 | ||
| NM_001273078.1 | NP_001260007.1 | 18 - 19 | 6376 | 85 | ||
| NM_001273079.1 | NP_001260008.1 | 5 - 6 | 1058 | 85 | ||
| NM_134962.3 | NP_608806.3 | 12 - 13 | 5150 | 85 | ||
More Information
Set Membership: |
|

CG43707
Genomic Map