Assay Details
| Gene Symbol: CG43707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273074.1 | NP_001260003.1 | 18 - 19 | 6540 | 65 | |
| NM_001273075.1 | NP_001260004.1 | 18 - 19 | 7102 | 65 | ||
| NM_001273076.1 | NP_001260005.1 | 20 - 21 | 6660 | 65 | ||
| NM_001273077.2 | NP_001260006.1 | 18 - 19 | 6540 | 65 | ||
| NM_001273078.1 | NP_001260007.1 | 19 - 20 | 6525 | 65 | ||
| NM_001273079.1 | NP_001260008.1 | 6 - 7 | 1207 | 65 | ||
| NM_134962.3 | NP_608806.3 | 13 - 14 | 5299 | 65 | ||
More Information
Set Membership: |
|

CG43707
Genomic Map