Assay Details
| Gene Symbol: CG43707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273074.1 | NP_001260003.1 | 10 - 11 | 3704 | 58 | |
| NM_001273075.1 | NP_001260004.1 | 10 - 11 | 4320 | 58 | ||
| NM_001273076.1 | NP_001260005.1 | 10 - 11 | 3668 | 58 | ||
| NM_001273077.2 | NP_001260006.1 | 10 - 11 | 3704 | 58 | ||
| NM_001273078.1 | NP_001260007.1 | 10 - 11 | 3668 | 58 | ||
| NM_134962.3 | NP_608806.3 | 5 - 6 | 2517 | 58 | ||
More Information
Set Membership: |
|

CG43707
Genomic Map