Assay Details
| Gene Symbol: CG43707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273074.1 | NP_001260003.1 | 11 - 12 | 3875 | 75 | |
| NM_001273077.2 | NP_001260006.1 | 11 - 12 | 3875 | 75 | ||
More Information
Set Membership: |
|

CG43707
Genomic Map