Assay Details
| Gene Symbol: TTLL4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273081.2 | NP_001260010.1 | 5 - 6 | 1895 | 72 | |
| NM_134966.3 | NP_608810.1 | 5 - 6 | 1814 | 72 | ||
| NM_164562.2 | NP_722946.1 | 5 - 6 | 1814 | 72 | ||
| GenBank mRNA | AY052082.1 | - | 5 - 6 | 1800 | 72 | |
More Information
Set Membership: |
|

TTLL4B
Genomic Map