Assay Details
| Gene Symbol: morgue | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134989.3 | NP_608833.1 | 1 - 2 | 223 | 67 | |
| GenBank mRNA | AF145603.1 | - | 1 - 2 | 150 | 67 | |
| AY078386.1 | - | 1 - 2 | 170 | 67 | ||
More Information
Set Membership: |
|

morgue
Genomic Map