Assay Details
| Gene Symbol: orb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274679.1 | NP_001261608.1 | 4 - 5 | 905 | 78 | |
| NM_001370022.1 | NP_001356977.1 | 3 - 4 | 915 | 78 | ||
| NM_001370023.1 | NP_001356978.1 | 3 - 4 | 915 | 78 | ||
| NM_140009.3 | NP_648266.1 | 3 - 4 | 915 | 78 | ||
| NM_168300.3 | NP_729427.1 | 3 - 4 | 915 | 78 | ||
| NM_168301.3 | NP_729428.1 | 3 - 4 | 842 | 78 | ||
| GenBank mRNA | AY052043.1 | - | 3 - 4 | 823 | 78 | |
| Gene Symbol: CG43783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274676.1 | NP_001261605.1 | 3 - 4 | 915 | 78 | |
| NM_001274677.1 | NP_001261606.1 | 3 - 4 | 839 | 78 | ||
More Information
Set Membership: |
|

orb2
Genomic Map