Assay Details
| Gene Symbol: orb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274679.1 | NP_001261608.1 | 5 - 6 | 973 | 63 | |
| NM_001370022.1 | NP_001356977.1 | 4 - 5 | 983 | 63 | ||
| NM_001370023.1 | NP_001356978.1 | 4 - 5 | 983 | 63 | ||
| NM_140009.3 | NP_648266.1 | 4 - 5 | 983 | 63 | ||
| NM_168300.3 | NP_729427.1 | 4 - 5 | 983 | 63 | ||
| NM_168301.3 | NP_729428.1 | 4 - 5 | 910 | 63 | ||
| GenBank mRNA | AY052043.1 | - | 4 - 5 | 891 | 63 | |
More Information
Set Membership: |
|

orb2
Genomic Map