Assay Details
| Gene Symbol: pix | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140015.3 | NP_648272.1 | 4 - 5 | 1645 | 96 | |
| NM_168306.2 | NP_729435.1 | 3 - 4 | 1606 | 96 | ||
| GenBank mRNA | AY089644.1 | - | 4 - 5 | 1634 | 96 | |
More Information
Set Membership: |
|

pix
Genomic Map