Assay Details
| Gene Symbol: CG13314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202144.1 | NP_001189073.1 | 2 | 431 | 72 | |
| NM_140018.2 | NP_648275.1 | 1 | 627 | 72 | ||
| GenBank mRNA | BT023648.1 | - | 1 | 627 | 72 | |
| Gene Symbol: Galk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168307.3 | NP_729438.1 | 6 | 1831 | 72 | |
More Information
Set Membership: |
|

CG13314
Genomic Map