Assay Details
| Gene Symbol: Galk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202145.1 | NP_001189074.1 | 3 - 4 | 601 | 66 | |
| NM_140019.4 | NP_648276.1 | 3 - 4 | 619 | 66 | ||
| NM_168307.3 | NP_729438.1 | 2 - 3 | 293 | 66 | ||
| NM_168308.3 | NP_729439.1 | 2 - 3 | 354 | 66 | ||
| GenBank mRNA | AY058337.1 | - | 3 - 4 | 593 | 66 | |
More Information
Set Membership: |
|

Galk
Genomic Map