Assay Details
| Gene Symbol: Tat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274699.1 | NP_001261628.1 | 5 - 6 | 968 | 84 | |
| NM_140052.2 | NP_648309.1 | 4 - 5 | 1010 | 84 | ||
| NM_140053.2 | NP_648310.1 | 5 - 6 | 1333 | 84 | ||
| NM_170630.2 | NP_729487.1 | 5 - 6 | 968 | 84 | ||
| NM_170631.1 | NP_729488.1 | 5 - 6 | 914 | 84 | ||
| NM_170632.2 | NP_729489.1 | 5 - 6 | 1414 | 84 | ||
| GenBank mRNA | AY061003.1 | - | 5 - 6 | 1320 | 84 | |
More Information
Set Membership: |
|

Tat
Genomic Map