Assay Details
| Gene Symbol: Zasp67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274725.1 | NP_001261654.1 | 1 - 2 | 265 | 122 | |
| NM_001274726.1 | NP_001261655.1 | 1 - 2 | 265 | 122 | ||
| NM_001300086.1 | NP_001287015.1 | 1 - 2 | 265 | 122 | ||
| NM_140101.2 | NP_648358.2 | 1 - 2 | 265 | 122 | ||
More Information
Set Membership: |
|

Zasp67
Genomic Map