Assay Details
| Gene Symbol: Zasp67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274726.1 | NP_001261655.1 | 7 - 8 | 2170 | 62 | |
| NM_140101.2 | NP_648358.2 | 7 - 8 | 2185 | 62 | ||
More Information
Set Membership: |
|

Zasp67
Genomic Map