Assay Details
| Gene Symbol: OXA1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140160.3 | NP_648417.1 | 2 - 3 | 363 | 71 | |
| NM_168424.2 | NP_729634.1 | 1 - 2 | 152 | 71 | ||
| GenBank mRNA | AF145596.1 | - | 1 - 2 | 393 | 71 | |
More Information
Set Membership: |
|

OXA1L
Genomic Map