Assay Details
| Gene Symbol: Plod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140194.2 | NP_648451.1 | 5 - 6 | 1290 | 106 | |
| NM_168452.2 | NP_729687.1 | 6 - 7 | 1204 | 106 | ||
More Information
Set Membership: |
|

Plod
Genomic Map