Assay Details
| Gene Symbol: Plod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140194.2 | NP_648451.1 | 8 - 9 | 1651 | 67 | |
| NM_168452.2 | NP_729687.1 | 9 - 10 | 1565 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plod| Gene Symbol: Plod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140194.2 | NP_648451.1 | 8 - 9 | 1651 | 67 | |
| NM_168452.2 | NP_729687.1 | 9 - 10 | 1565 | 67 | ||
Set Membership: |
|