Assay Details
| Gene Symbol: Plod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140194.2 | NP_648451.1 | 9 - 10 | 2089 | 66 | |
| NM_168452.2 | NP_729687.1 | 10 - 11 | 2003 | 66 | ||
More Information
Set Membership: |
|

Plod
Genomic Map