Assay Details
| Gene Symbol: chrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300105.1 | NP_001287034.1 | 2 - 3 | 760 | 79 | |
| NM_140213.5 | NP_648470.2 | 2 - 3 | 1697 | 79 | ||
| GenBank mRNA | AY060367.1 | - | 2 - 3 | 1414 | 79 | |
More Information
Set Membership: |
|

chrb
Genomic Map