Assay Details
| Gene Symbol: chrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300105.1 | NP_001287034.1 | 3 - 4 | 875 | 63 | |
| NM_140213.5 | NP_648470.2 | 3 - 4 | 1812 | 63 | ||
| GenBank mRNA | AY060367.1 | - | 3 - 4 | 1529 | 63 | |
More Information
Set Membership: |
|

chrb
Genomic Map