Assay Details
| Gene Symbol: Dph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274793.1 | NP_001261722.1 | 2 - 3 | 849 | 95 | |
| NM_140239.4 | NP_648496.1 | 2 - 3 | 854 | 95 | ||
| NM_168471.3 | NP_729735.1 | 2 - 3 | 913 | 95 | ||
| GenBank mRNA | AY069411.1 | - | 2 - 3 | 860 | 95 | |
More Information
Set Membership: |
|

Dph1
Genomic Map