Assay Details
| Gene Symbol: Bmcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140244.5 | NP_648501.1 | 2 - 3 | 403 | 107 | |
| NM_168472.2 | NP_729738.1 | 2 - 3 | 403 | 107 | ||
| GenBank mRNA | AY051433.1 | - | 2 - 3 | 365 | 107 | |
More Information
Set Membership: |
|

Bmcp
Genomic Map