Assay Details
| Gene Symbol: Bmcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140244.5 | NP_648501.1 | 5 - 6 | 1446 | 162 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmcp| Gene Symbol: Bmcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140244.5 | NP_648501.1 | 5 - 6 | 1446 | 162 | |
Set Membership: |
|