Assay Details
| Gene Symbol: Rpt4R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300110.1 | NP_001287039.1 | 1 - 2 | 808 | 64 | |
| NM_140268.2 | NP_648525.1 | 1 - 2 | 697 | 64 | ||
More Information
Set Membership: |
|

Rpt4R
Genomic Map