Assay Details
| Gene Symbol: Pop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169958.2 | NP_001163429.1 | 1 - 2 | 416 | 63 | |
| NM_001300112.1 | NP_001287041.1 | 2 - 3 | 677 | 63 | ||
| NM_001382181.1 | NP_001369048.1 | 2 - 3 | 677 | 63 | ||
| NM_140281.3 | NP_648538.1 | 2 - 3 | 677 | 63 | ||
| NM_168484.2 | NP_729775.1 | 3 - 4 | 882 | 63 | ||
| NM_168485.2 | NP_729776.1 | 3 - 4 | 816 | 63 | ||
| GenBank mRNA | AY047553.1 | - | 2 - 3 | 628 | 63 | |
| BT001837.1 | - | 3 - 4 | 775 | 63 | ||
More Information
Set Membership: |
|

Pop2
Genomic Map