Assay Details
| Gene Symbol: CrzR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274826.1 | NP_001261755.1 | 3 - 4 | 1492 | 67 | |
| NM_140314.4 | NP_648571.1 | 3 - 4 | 1492 | 67 | ||
| GenBank mRNA | AY071408.1 | - | 3 - 4 | 1484 | 67 | |
More Information
Set Membership: |
|

CrzR
Genomic Map