Assay Details
| Gene Symbol: Smyd4-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140317.3 | NP_648574.1 | 2 - 3 | 1033 | 71 | |
| GenBank mRNA | AY071300.1 | - | 2 - 3 | 920 | 71 | |
More Information
Set Membership: |
|

Smyd4-2
Genomic Map