Assay Details
| Gene Symbol: CG42709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169963.2 | NP_001163434.1 | 4 - 5 | 697 | 87 | |
| NM_001300122.1 | NP_001287051.1 | 4 - 5 | 697 | 87 | ||
| NM_140343.4 | NP_648600.1 | 4 - 5 | 830 | 87 | ||
| NM_168516.3 | NP_729833.1 | 3 - 4 | 856 | 87 | ||
| GenBank mRNA | AY058716.1 | - | 4 - 5 | 809 | 87 | |
More Information
Set Membership: |
|

CG42709
Genomic Map