Assay Details
| Gene Symbol: Atg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169962.2 | NP_001163433.1 | 10 - 11 | 3161 | 72 | |
| NM_140344.3 | NP_648601.1 | 11 - 12 | 3101 | 72 | ||
| GenBank mRNA | AY129445.1 | - | 10 - 11 | 3130 | 72 | |
| BT021340.1 | - | 11 - 12 | 3036 | 72 | ||
More Information
Set Membership: |
|

Atg1
Genomic Map