Assay Details
| Gene Symbol: ste14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274864.1 | NP_001261793.1 | 1 - 2 | 354 | 61 | |
| NM_140366.3 | NP_648623.1 | 1 - 2 | 354 | 61 | ||
| GenBank mRNA | AY070700.1 | - | 1 - 2 | 370 | 61 | |
| AY071003.1 | - | 1 - 2 | 342 | 61 | ||
More Information
Set Membership: |
|

ste14
Genomic Map